missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuropilin-1 Antibody (761705) [Janelia Fluor™ 525], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5994JF525
This item is not returnable.
View return policy
Description
Neuropilin-1 Monoclonal antibody specifically detects Neuropilin-1 in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Neuropilin-1 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| BDCA4, CD304, CD304 antigen, DKFZp686A03134, DKFZp781F1414, neuropilin 1, neuropilin-1, NP1, NRP, transmembrane receptor, Vascular endothelial cell growth factor 165 receptor, VEGF165R | |
| Mouse myeloma cell line NS0-derived recombinant mouse Neuropilin-1, Phe21-Pro856 , Accession # P97333 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 761705 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, Breast Cancer, Cancer, Cardiovascular Biology, Neuroscience, Phospho Specific, Virology Bacteria and Parasites | |
| 8829 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction