missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neuropilin-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595832
This item is not returnable.
View return policy
Description
Positive Samples: SH-SY5Y, A-549, U-87MG, 293T, mouse brain; Cellular Location: Cell membrane, Single-pass type I membrane protein Immunogen sequence: FELVDGVKLK KDKLNTQSTY SEA.
Neuropilin 1 is a 923 amino acid containing membrane bound coreceptor to a tyrosine kinase receptor belonging to the neuropilin family with two CUB domains, two F5/8 type C domains and a MAM domain. Neuropilin 1 forms a heterodimer with NRP2 (Neuropilin 2) and is known to bind to plexin (PLXNB1) to form semaphorin receptors. Expressed mostly in the nervous system this receptor is implicated in a wide range of functions including in the development of the cardiovascular system, angiogenesis, repulsive axon guidance, formation of certain neuronal circuits, cell survival, control of neuronal migration and organogenesis outside the nervous system. This semaphorin receptor functions as receptors for both the semaphorins and vascular endothelial growth factor and is also a regulator of VEGF-induced angiogenesis. Neuropilin 1 may be involved in aggressive advanced prostate carcinoma and in the progression of breast cancer.
Specifications
| Neuropilin-1 | |
| Polyclonal | |
| Unconjugated | |
| NRP1 | |
| A5 protein; BDCA4; C530029I03; CD304; Neuropilin; neuropilin 1; Neuropilin1; neuropilin-1; Neuropilin-1 precursor (A5 protein); NP1; NP-1; Npn1; NPN-1; NRP; NRP 1; Nrp1; sNRP 1; sNRP1; soluble NRP 1; soluble NRP1; transmembrane receptor; vascular endothelial cell growth factor 165 receptor; VEGF165R | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18186, 246331, 8829 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.75 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14786, P97333, Q9QWJ9 | |
| NRP1 | |
| A synthetic peptide corresponding to a sequence within amino acids 824-923 of human Neuropilin-1 (NRP1) (NP_0038645). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction