missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuroplastin Antibody (OTI3C4), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72966AF532
This item is not returnable.
View return policy
Description
Neuroplastin Monoclonal specifically detects Neuroplastin in Human, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Neuroplastin | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| NPTN | |
| Full length recombinant protein of human Neuroplastin (NP_059429) produced in HEK293T cells. | |
| 0.1 mL | |
| 27020.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3C4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| GP55Stromal cell-derived receptor 1, GP65SDR-1, MGC102805, neuroplastin, np55, np65, SDFR1DKFZp686L2477, SDR1stromal cell derived factor receptor 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Rat, Canine, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction