missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuroplastin Antibody (OTI3C4), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72966MFV610
This item is not returnable.
View return policy
Description
Neuroplastin Monoclonal antibody specifically detects Neuroplastin in Human, Rat, Canine, Monkey samples. It is validated for Flow Cytometry
Specifications
| Neuroplastin | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 27020 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI3C4 | |
| Flow Cytometry | |
| GP55Stromal cell-derived receptor 1, GP65SDR-1, MGC102805, neuroplastin, np55, np65, SDFR1DKFZp686L2477, SDR1stromal cell derived factor receptor 1 | |
| Full length human recombinant protein of human NPTN(NP_059429) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Rat, Canine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction