missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurotrimin/HNT Antibody (MM0485-5X25), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11813AF488
This item is not returnable.
View return policy
Description
Neurotrimin/HNT Monoclonal specifically detects Neurotrimin/HNT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Neurotrimin/HNT | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NTM | |
| Human recombinant Neurotrimin/HNT | |
| 0.1 mL | |
| 50863 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0485-5X25 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| hNT, IgLON family member 2, IGLON2HNTNT, MGC60329, neurotrimin, NTRI | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction