Learn More
Neurotrophin 4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12297-1-AP-150UL
 View more versions of this product
Product Code. 16809933
  • € 410.00 / 150µL
Description
This gene is a member of a family of neurotrophic factors, neurotrophins, that control survival and differentiation of mammalian neurons. The expression of this gene is ubiquitous and less influenced by environmental signals. While knock-outs of other neurotrophins including nerve growth factor, brain-derived neurotrophic factor, and neurotrophin 3 prove lethal during early postnatal development, NTF5-deficient mice only show minor cellular deficits and develop normally to adulthood.
Specifications
Specifications
Neurotrophin 4
Polyclonal
Unconjugated
NTF4
neurotrophin 4, Neurotrophin 5, Neutrophic factor 4, NT 4, NT 4/5, NT 5, NT4, NT5, NTF4, NTF5
Rabbit
Antigen Affinity Chromatography
RUO
25730, 4909, 78405
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P34130, P34131, Q80VU4
Ntf4
Neurotrophin 4 Fusion Protein Ag2938
150 μL
Primary
Human, Mouse, Rat