Learn More
DiscoverX Neurturin
Click to view available options
:
20 μg
Description
Neurturin is a protein member of the TGF-beta family. Functions to support sympathetic neuron survival, and may be involved in the development and maintenance of the CNS as well as the control of the hemopoietic cell population. Has been tested in clinical trials for the treatment of nerodegenerative diseases like Huntington's and Parkinson's disease. Acts as an agonist for c-Ret. Provided as 20μg of dry powder alongside reconstitution buffer.
Specifications
Specifications
| Quantity | 20 μg |
| Product Type | Control Ligands |