Learn More
DiscoverX Neurturin
Product Code. 17342469
Encompass
Click to view available options
:
20 μg
Product Code. 17342469
Brand: DiscoverX 921141
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Neurturin is an agonist for c-Ret. Provided as 20μg of dry powder alongside reconstitution buffer.
Neurturin is a protein member of the TGF-beta family. Functions to support sympathetic neuron survival, and may be involved in the development and maintenance of the CNS as well as the control of the hemopoietic cell population. Has been tested in clinical trials for the treatment of nerodegenerative diseases like Huntington's and Parkinson's disease. Acts as an agonist for c-Ret. Provided as 20μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 20 μg
Product Type Control Ligands