missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeutraControl TGF beta 1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 69512-1-IG-100UG
This item is not returnable.
View return policy
Description
The NeutraControl TGF beta 1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraControl TGF beta 1. This antibody recognizes human antigen. The NeutraControl TGF beta 1 antibody has been validated for the following applications: ELISA, Non-Neutralization analysis.Specifications
| TGF beta 1 | |
| Monoclonal | |
| 1000 μg/mL | |
| CED; DPD1; TGFB; TGFbeta | |
| RecombinantProtein | |
| 100 μg | |
| Primary | |
| Human | |
| Neu Control | |
| IgG1 |
| ELISA, Non-neutralization | |
| 1D3D11 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 7040 | |
| Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering