missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeutraKine™ BMP-7 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 69011-1-IG-100UG
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The NeutraKine™ BMP-7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine™ BMP-7. This antibody recognizes human, mouse antigen. The NeutraKine™ BMP-7 antibody has been validated for the following applications: FC (Intra), Neutralization, ELISA analysis.Spezifikation
| BMP-7 | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 655 | |
| Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. | |
| Liquid |
| Flow Cytometry (Intracellular Staining), Neutralization, ELISA | |
| 3E9G7 | |
| OP-1 | |
| RecombinantProtein | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Neu Antibody | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur