Learn More
NeutraKine™ BMP-7 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 69011-1-IG-25UG
 View more versions of this product
Product Code. 30238002
  • € 149.97 / 25µg
Description
The NeutraKine™ BMP-7 antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine™ BMP-7. This antibody recognizes human, mouse antigen. The NeutraKine™ BMP-7 antibody has been validated for the following applications: FC (Intra), Neutralization, ELISA analysis.
Specifications
Specifications
BMP-7
Monoclonal
Unconjugated
Mouse
Protein G purification
RUO
655
Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Liquid
Flow Cytometry (Intracellular Staining), Neutralization, ELISA
3E9G7
OP-1
RecombinantProtein
25 μg
Primary
Human, Mouse
Neu Antibody