missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 405 Anti-Human CD29 (TS2/16), Proteintech

Product Code. 30236318 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30236318 100 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30236318 Supplier Proteintech Supplier No. CL40565191100TESTS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CD29 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD29. This antibody recognizes human antigen. The CD29 antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 405.

Specifications

Antigen CD29
Applications Flow Cytometry
Classification Monoclonal
Clone TS2/16
Concentration 5 μl/test
Conjugate CoraLite Plus 405
Gene Symbols CD29; FNRB; GPIIA; MDF2; MSK12; VLA-BETA; VLAB
Host Species Mouse
Purification Method Affinity purification
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3688
Target Species Human
Content And Storage Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment.
Product Type Flow Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.