Learn More
NeutraKine FLT3 Ligand Mouse, anti-Human, Clone: 3E1A3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 69032-1-IG-100UG
 View more versions of this product
Product Code. 18095145
  • € 319.00 / 100µg
Description
The NeutraKine FLT3 Ligand antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine FLT3 Ligand. This antibody recognizes human antigen. The NeutraKine FLT3 Ligand antibody has been validated for the following applications: Neutralization, ELISA analysis.
Specifications
Specifications
NeutraKine FLT3 Ligand
3E1A3
Mouse
Primary
Human
150 kDa
IgG1
ELISA, Neutralization
FL, Flt3 ligand, Flt3L, FLT3LG, SL cytokine
100 μg
2323
Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.