Learn More
NeutraKine™IL-9 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 69031-1-IG-100UG
 View more versions of this product
Product Code. 30233571
  • € 321.08 / 100µg
Description
The NeutraKine™IL-9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinantprotein of human NeutraKine™IL-9. This antibody recognizes human antigen. The NeutraKine™IL-9 antibody has been validated for the following applications: ELISA, Neutralization analysis.
Specifications
Specifications
IL-9
Monoclonal
Unconjugated
Mouse
Protein A purification
RUO
3578
Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20°C) and (-80°C). Upon reconstitution we recommend that the solution can be stored at(4°C) for short term or at(-20°C) to (-80°C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.
Liquid
ELISA, Neutralization
1B3A10
HP40; IL-9; P40
RecombinantProtein
100 μg
Primary
Human
Neu Antibody
IgG2a