Learn More
Honeywell™ Neutron Multiple-Use Earplugs
Product Code. 12603453
Change view
Click to view available options
:
Translucent blue, corded
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12603453 Translucent blue, corded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12603453 Supplier Honeywell™ Supplier No. 1029810
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Helps prevent exposure to harmful noise and allows you to hear colleagues and customers without removing them.
  • Helps prevent exposure to harmful noise and allows you to hear colleagues and customers
  • Keeps you better connected and protected in moderate noise level workplaces
  • Optimal attenuation across all frequencies
  • Allows you to clearly hear voices, signals, alarms without removing earplugs
  • Ideal for moderate noise environments such as pharmaceutical production, transportation, utilities, general manufacturing, entertainment venues
  • Visible, yet discreet, detachable cord
  • Streamlined, long flange sits comfortably inside the ear canal for a more comfortable in-ear seal and secure fit
  • li>One-year guarantee against defects in material and workmanship: toes not cover damage caused by misuse, abuse or unauthorized modifications
TRUSTED_SUSTAINABILITY
Specifications
Color Translucent Blue
Certifications/Compliance (Europe) 2003/10/EC - EU Hearing Directive, 89/686/EEC - European Directive/PPE, Category II - EC Category PPE, EN 352-2:1993 - EU Standards
Noise Reduction Rating (NRR) SNR: 20 dB
Plug Shape Dual-Flange
Product Type Corded, re-usable ear plugs
Material TPE