missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neutrophil Elastase/ELA2 Antibody (39A), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64392V
This item is not returnable.
View return policy
Description
Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Flow Cytometry, ELISA.
Specifications
| Elastase | |
| Monoclonal | |
| DyLight 405 | |
| P08246 | |
| ELANE | |
| Elastase from human neutrophils | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 23436 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 39A | |
| Flow Cytometry, ELISA | |
| CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-64392 is specific for human neutrophil elastase. | |
| Store at 4C in the dark. | |
| IgG1 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido