missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neutrophil Elastase/ELA2 Antibody (39A), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64392G
This item is not returnable.
View return policy
Description
Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for Flow Cytometry, ELISA.
Specifications
| Elastase | |
| Monoclonal | |
| DyLight 488 | |
| P08246 | |
| ELANE | |
| Elastase from human neutrophils | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 23436 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 39A | |
| Flow Cytometry, ELISA | |
| CBPP, cholesterol-binding pancreatic protease, chymotrypsin-like elastase family member 3B, chymotrypsin-like elastase family, member 3B, E1, EC 3.4.21, EC 3.4.21.70, EL-1, ELA3B, elastase 1, elastase 3B, pancreatic, Elastase IIIB, elastase-3B, fecal elastase 1, pancreatic elastase 1, pancreatic endopeptidase E, Protease E, proteinase E | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-64392 is specific for human neutrophil elastase. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction