Learn More
Neutrophil Elastase/ELA2 Antibody (950312) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB91673Y
Product Code. 18523538
  • € 634.00 / 0.10mL
Description
Neutrophil Elastase/ELA2 Monoclonal antibody specifically detects Neutrophil Elastase/ELA2 in Human samples. It is validated for ELISA, Flow Cytometry (Intracellular Staining), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Neutrophil Elastase/ELA2
Monoclonal
DyLight 680
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
ELISA, Flow Cytometry (Intracellular Staining), CyTOF
950312
ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready
Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1
Chinese hamster ovary cell line CHO-derived recombinant human Elastase/ELA-2, Met1-Asn252, Accession # P08246
0.1 mL
Cancer, Neuroscience
1991
Store at 4°C in the dark.