missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Neutrophil elastase Recombinant Rabbit Monoclonal Antibody (2H12)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA549834
This item is not returnable.
View return policy
Description
Neutrophil elastase Recombinant Monoclonal Antibody for IHC (P), Flow, ELISA
The neutrophil form of elastase is 218 amino acids long, with two asparagine-linked carbohydrate chains (see glycosylation). It is present in azurophil granules in the neutrophil cytoplasm. There appear to be two forms of neutrophil elastase, termed IIa and IIb.
Specifications
| Neutrophil elastase | |
| Recombinant Monoclonal | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P08246 | |
| ELANE | |
| A synthesized peptide derived from human Neutrophil Elastase. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 2H12 | |
| Unconjugated | |
| ELANE | |
| bone marrow serine protease; ELA2; ELA2A; ELANE; Elastase; elastase 2, neutrophil; elastase, neutrophil expressed; Elastase2; elastase-2; F430011M15Rik; GE; granulocyte-derived elastase; HLE; HNE; Human leukocyte elastase; Leukocyte elastase; medullasin; NE; neutrophil elastase; PE-1; PMN elastase; PMN-E; polymorphonuclear elastase; SCN1; serine protease | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1991 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction