Learn More
Neutrophils Antibody (RM0028-3G23), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP2-12439CL1
Product Code. 30140635
  • € 1100.00 / 0.10mL
Description
Neutrophils Monoclonal antibody specifically detects Neutrophils in Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Neutrophils
Monoclonal
CoraFluor 1
Rat
Protein G purified
RUO
Mouse
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
RM0028-3G23
PBS
Isolated mouse neutrophil cells
0.1 mL
Primary
Store at 4°C in the dark. Do not freeze.
IgG2a