Learn More
Eppendorf™ Gable Covers for Model C76 Water Bath Shaker
Product Code. 11379874
Change view
Click to view available options
Material:
Plexiglass™
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
11379874 Plexiglass™ Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11379874 Supplier Eppendorf™ Supplier No. M12312010
Only null left
Add to basket
Add to basket
For C76 Water Bath Shaker
  • Plexiglas™ cover permits temperatures up to 60°C Stainless-steel cover required for temperatures above 60°C
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Model C76 water bath shaker
Material Plexiglass™