missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Newcastle Disease Virus (NDV) Antibody (9F7), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
€ 553.00
Specifications
| Antigen | Newcastle Disease Virus (NDV) |
|---|---|
| Clone | 9F7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Janelia Fluor 646 |
Description
Newcastle Disease Virus (NDV) Monoclonal antibody specifically detects Newcastle Disease Virus (NDV) in Virus samples. It is validated for Western Blot, Haemagglutination, ELISASpecifications
| Newcastle Disease Virus (NDV) | |
| Western Blot | |
| Janelia Fluor 646 | |
| Mouse | |
| Virus | |
| NDV | |
| IgG1 | |
| Protein G purified |
| 9F7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Sodium Borate | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Newcastle disease virus (La-sota strain). | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title