missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-H Antibody (SPM203), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47833CL1
This item is not returnable.
View return policy
Description
NF-H Monoclonal antibody specifically detects NF-H in Chicken,Human,Mouse,Porcine,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),CyTOF
Specifications
| NF-H | |
| Monoclonal | |
| CoraFluor 1 | |
| 200 kDa neurofilament protein, KIAA0845, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H | |
| Triton-X 100 insoluble protein fraction of rat brain (Uniprot: P12036) | |
| 0.1 mL | |
| Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
| 4744 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), CyTOF | |
| SPM203 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Chicken, Human, Mouse, Pig, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction