missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NF-H Monoclonal Antibody (AH1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547473
This item is not returnable.
View return policy
Description
NF-H Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
Specifications
| NF-H | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5mM sodium azide | |
| P12036, P12037, P16884, P19246 | |
| Nefh | |
| Native NF-H purified from bovine spinal cord. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine, Pig, Horse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| AH1 | |
| Unconjugated | |
| Nefh | |
| 200 kDa neurofilament protein; CMT2CC; heavy neurofilament protein; I79_012512; intermediate filament protein; KIAA0845; LOW QUALITY PROTEIN: neurofilament heavy polypeptide; mKIAA0845; NEF3; Nefh; nefh.L; neurofilament; neurofilament 200kDa; neurofilament heavy; neurofilament heavy L homeolog; neurofilament heavy polypeptide; neurofilament heavy polypeptide-like; neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, heavy polypeptide L homeolog; neurofilament-H; NF 200 kD; NF200; Nfh; NF-H; NHC; XELAEV_18007418mg | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100064328, 100156492, 24587, 380684, 4744, 528842 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction