missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NF-H Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5143557
This item is not returnable.
View return policy
Description
NF-H Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IHC (PFA)
Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
Specifications
| NF-H | |
| Polyclonal | |
| Unconjugated | |
| Nefh | |
| 200 kDa neurofilament protein; CMT2CC; heavy neurofilament protein; I79_012512; intermediate filament protein; KIAA0845; LOW QUALITY PROTEIN: neurofilament heavy polypeptide; mKIAA0845; NEF3; Nefh; nefh.L; neurofilament; neurofilament 200kDa; neurofilament heavy; neurofilament heavy L homeolog; neurofilament heavy polypeptide; neurofilament heavy polypeptide-like; neurofilament triplet H protein; neurofilament, heavy polypeptide; neurofilament, heavy polypeptide 200kDa; neurofilament, heavy polypeptide L homeolog; neurofilament-H; NF 200 kD; NF200; Nfh; NF-H; NHC; XELAEV_18007418mg | |
| Goat | |
| Affinity chromatography | |
| RUO | |
| 100064328, 100156492, 24587, 380684, 4744, 528842 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 5mM sodium azide | |
| P12036, P12037, P16884, P19246 | |
| Nefh | |
| Native NF-H purified from bovine spinal cord. | |
| 500 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine, Pig, Horse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction