Learn More
NF-L Antibody (NR-4), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals NBP2-47969AF647
Code produit. 18779770
  • € 577.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
NF-L Monoclonal antibody specifically detects NF-L in Human, Rat, Porcine, Bovine, Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
TRUSTED_SUSTAINABILITY
Spécification
Spécification
NF-L
Monoclonal
Alexa Fluor 647
68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa
Crude neurofilament preparation from porcine spinal cord
0.1 mL
Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience
4747
Store at 4°C in the dark.
IgG1 κ
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
NR-4
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Rat, Pig, Bovine, Chicken