missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-L Antibody (SPM204) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-48293
This item is not returnable.
View return policy
Description
NF-L Monoclonal specifically detects NF-L in Human, Rat, Porcine, Bovine, Chicken samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NF-L | |
| Monoclonal | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| NEFL | |
| Neurofilament purified from human brain | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This monoclonal antibody reacts with a 68kDa protein, identified as light sub-unit of neurofilaments (NF-L). Neurofilaments make up the main structural elements of axons and dendrites and are found in neurons, peripheral nerves, and sympathetic ganglion cells. Neurofilaments consist of three major subunits with molecular weights of 68kDa (NF-L), 160kDa (NF-M) and 200kDa (NF-H). Anti-neurofilament stains a number of neural, neuroendocrine, and endocrine tumors. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas, and neuroblastomas stain positively for anti-neurofilament. Neurofilaments are also present in paragangliomas as well as adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilament. | |
| Store at 4C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| SPM204 | |
| Immunohistochemistry-Paraffin | |
| 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa | |
| Mouse | |
| 68 kDa | |
| 7 mL | |
| Autophagy, Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience | |
| 4747 | |
| Human, Rat, Pig, Chicken, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction