missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF2/Merlin Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99421AF700
This item is not returnable.
View return policy
Description
NF2/Merlin Polyclonal antibody specifically detects NF2/Merlin in Mouse samples. It is validated for Western Blot
Specifications
| NF2/Merlin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| ACN, BANF, merlin, moesin-ezrin-radixin like, Moesin-ezrin-radixin-like protein, moesin-ezrin-radizin-like protein, neurofibromin 2 (bilateral acoustic neuroma), neurofibromin 2 (merlin), neurofibromin-2, SCH, Schwannomerlin, Schwannomin | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NF2/Merlin | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Tumor Suppressors | |
| 4771 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction