Learn More
Invitrogen™ NFAT5 Monoclonal Antibody (K49018_1C5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555769
Description
Sequence of this protein is as follows: GQYPVKSEGK ELKIVVQPET QHRARYLTEG SRGSVKDRTQ QGFPTVKLEG HNEPVVLQVF VGNDSGRVKP HGFYQACRVT GRNTTPCKEV DIEGTTVIEV GLDPSNNMTL AVDCVGILKL RNADVEARIG IAGSKKKSTR ARLVFRVNIM RKDGSTLTLQ TPSSPILCTQ PAGVPEILKK SLHSCSVKGE EEVFLIGKNF LKGTKVIFQE NVSDENSWKS EAEIDMELFH QNHLIVKVPP YHDQHITLPV SVGIYVVTNA GRSHDVQPFT
The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.
Specifications
| NFAT5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O94916 | |
| NFAT5 | |
| Recombinant human NFAT5 (immunogen range is 271-540). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| K49018_1C5 | |
| Unconjugated | |
| NFAT5 | |
| AI225870; B130038B15Rik; CAG-8; CAG80; glutamine rich protein H65; KIAA0827; mKIAA0827; Nfat; Nfat5; NF-AT5; NFATL1; NFAT-like protein 1; nfatz; nuclear factor of activated T cells 5; nuclear factor of activated T-cells 5; nuclear factor of activated T-cells 5, tonicity-responsive; OREBP; osmotic response element-binding protein; rel domain-containing transcription factor NFAT5; T-cell transcription factor NFAT5; TonE-binding protein; TONEBP; tonicity-responsive enhancer binding protein; tonicity-responsive enhancer-binding protein | |
| Mouse | |
| Protein A | |
| RUO | |
| 10725 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.