Learn More
NFATC1 Mouse anti-Human, Unconjugated, Clone: 1E1B10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66963-1-IG-150UL
This item is not returnable.
View return policy
Description
The NFATC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NFATC1. This antibody recognizes human antigen. The NFATC1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| NFATC1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 1E1B10 | |
| Unconjugated | |
| 150 μL | |
| 4772 | |
| IgG1 |