missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFATC2IP, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00084901-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MSEPLQSVVDHMATHLGVSPSRILLLFGETELSPTATPRTLKLGVADIIDCVVLTSSPEATETSQQLQLRVQGKEKHQTLEVSLSRDSPLKTLMSHYEEAMGLSGRKLSFFFDGTKLSGRELPADLGMESGDLIEVWGSpecifications
| NFATC2IP | |
| Polyclonal | |
| No additive | |
| BC021551 | |
| NFATC2IP | |
| NFATC2IP (AAH21551.1, 1 a.a. ∼ 138 a.a) full-length human protein. | |
| RUO | |
| 84901 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2 interacting protein | |
| FLJ14639/MGC126790/MGC138387 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction