missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFATC3/NFAT4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58079
This item is not returnable.
View return policy
Description
NFATC3/NFAT4 Polyclonal specifically detects NFATC3/NFAT4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| NFATC3/NFAT4 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| NF-AT4, NFAT4nuclear factor of activated T-cells, cytoplasmic 3, NFATc3, NF-ATc3, NFATx, nuclear factor of activated T-cells c3 isoform IE-Xa, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 3, T cell transcription factor NFAT4, T-cell transcription factor NFAT4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| NFATC3 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SHLPQLQCRDESVSKEQHMIPSPIVHQPFQVTPTPPVGSSYQPMQTNVVYNGPTCLPINAASSQEFDSVLFQQDATLSGLVNLGCQPLSSIPFHSSNSGSTGHLLAHTPHSVHTLPHLQSMGYHCSNTGQRSLSSPVADQITGQP | |
| 100 μL | |
| Adaptive Immunity, Immunology, Wnt Signaling Pathway | |
| 4775 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction