missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NFATC3 Polyclonal Antibody

Product Code. 15527521
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15527521 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15527521 Supplier Invitrogen™ Supplier No. PA536101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 115 kDa. Purity is >95% by SDS-PAGE.

This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein is located only in the cytoplasm. When translation ends, it interacts with the protein that is a functional homolog of yeast Upf2p to trigger mRNA decapping. Use of multiple polyadenylation sites has been noted for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFATC3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene NFATC3
Gene Accession No. P97305, Q12968
Gene Alias C80703; D8Ertd281e; NFAC3; NFAT4; NF-AT4; NFATC3; NF-ATc3; NFATx; NFC3; nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 3; nuclear factor of activated T-cells 3; nuclear factor of activated T-cells c3 isoform IE-Xa; nuclear factor of activated T-cells, cytoplasmic 3; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 3; T cell transcription factor NFAT4; T-cell transcription factor NFAT4
Gene Symbols NFATC3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 130-180 of human NFAT4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18021, 361400, 4775
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.