missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NFATC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585559
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (91%), Rat (91%). Positive Control: NFATC4-transfected 293T cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This protein is an adapter or scaffold protein, which participates in the assembly of numerous protein complexes and is involved in several cellular processes such as cell fate determination, cytoskeletal organization, repression of gene transcription, mitosis, cell-cell adhesion, cell differentiation, proliferation and migration.
Specifications
| NFATC4 | |
| Polyclonal | |
| Unconjugated | |
| Nfatc4 | |
| 3110041H08Rik; AW107667; AW546455; Nfat3; NF-AT3; Nfatc4; NF-ATc4; nuclear factor of activated T cells, cytoplasmic, calcineurin dependent 4; nuclear factor of activated T-cells 4; nuclear factor of activated T-cells, cytoplasmic 4; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4; T-cell transcription factor NFAT3; transcription complex subunit NF-ATc4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4776 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q14934 | |
| Nfatc4 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human NFATC4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction