missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFE2L2, Mouse anti-Human, Clone: 2D5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00004780-M05.100ug
This item is not returnable.
View return policy
Description
Sequence: SPSVASPEHSVESSSYGDTLLGLSDSEVEELDSAPGSVKQNGPKTPVHSSGDMVQPLSPSQGQSTHVHDAQCENTPEKELPVSPGHRKTPFTKDKHSSRLSpecifications
| NFE2L2 | |
| Monoclonal | |
| Unconjugated | |
| nuclear factor (erythroid-derived 2)-like 2 | |
| NRF2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2D5 | |
| In 1x PBS, pH 7.4 | |
| NM_006164 | |
| NFE2L2 | |
| NFE2L2 (NP_006155.2, 351 a.a. ∼ 450 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 4780 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction