missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NFE4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA549450

Product Code. 15977434

  • € 530.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NFE4 Polyclonal Antibody for Western Blot

Functions as part of the SSP (stage selector protein) complex, a complex that contributes to the preferential expression of the gamma-gene in fetal erythroid cells by facilitating the interaction of the gamma-globin genes with enhancer elements contained in the locus control region (LCR). The complex binds to the stage selector element (SSE) in the proximal gamma-globin promoter. In contrast, isoform 2 acts as a repressor of gamma- globin gene expression by preventing NFE2 and RNA polymerase II recruitment to the promoter.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NFE4
Polyclonal
Unconjugated
NFE4
fetal globin activator NF-E4; gamma-globin gene activator; NFE4; NF-E4; nuclear factor, erythroid 4; transcription factor NF-E4
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
58160
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.45 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q86UQ8
NFE4
KLH conjugated synthetic peptide between 135-169 amino acids from the C-terminal region of human NFE4.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.