missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFI Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA536188
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 47 kDa (NFIB) and 56 kDa (NFIA, NFIC, NFIX). Purity is >95% by SDS-PAGE.
NFI Polyclonal antibody specifically detects NFI in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Specifications
| NFI | |
| Polyclonal | |
| Unconjugated | |
| NFIA | |
| 1110019L22Rik; 1110047K16Rik; 1500041O16Rik; 6720429L07Rik; 9430022M17Rik; AA589446; AI746521; CCAAT-binding transcription factor; CCAAT-box-binding transcription factor; CTF; CTF5; E030026I10Rik; HMGIC/NFIB; KIAA1439; MRSHSS; NF1A; NF1-A; NF1-B; NF1-C; Nf1l21; NF1-X; NF1-X1 protein; NFI; NF-I; NF-I/A; NF-I/B; NF-I/C; NF-I/X; NFIA; NFI-A; Nfib; NFI-B; NFIB2; NFIB3; Nfic; NFI-C; NFI-L; NFI-RED; Nfix; NFI-X; Nuclear factor 1 A-type; nuclear factor 1 B-type; Nuclear factor 1 C-type; nuclear factor 1 X-type; nuclear factor 1/A; nuclear factor 1/B; nuclear factor 1/C; nuclear factor 1/X; nuclear factor 1-C2; nuclear factor I A; nuclear factor I B; nuclear factor I C; nuclear factor I X; nuclear factor I/A; Nuclear factor I/B; nuclear factor I/C; nuclear factor I/C (CCAAT-binding transcription factor); nuclear factor I/X; nuclear factor I/X (CCAAT-binding transcription factor); olfactory epithelium nuclear factor I-B; olfactory epithelium nuclear factor I-C; SOTOS2; TGGCA-binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18027, 18028, 18029, 18032, 25492, 29227, 29228, 4774, 4781, 4782, 4784, 81524 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.2 | |
| O00712, P08651, P09414, P70255, P70257, P97863, Q02780, Q12857, Q14938 | |
| NFIA, NFIB, Nfic, NFIX | |
| Synthetic peptide corresponding to the amino acids 20-60 of human NF-1A | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction