missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFIA Monoclonal Antibody (OTI4G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811215S
This item is not returnable.
View return policy
Description
NFIA Monoclonal antibody specifically detects NFIA in Human samples. It is validated for Western BlotSpecifications
| NFIA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BZP6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 27159 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4G8 | |
| Unconjugated | |
| CHIA | |
| 1110047K16Rik; 9430022M17Rik; CCAAT-box-binding transcription factor; CTF; KIAA1439; NF1A; NF1-A; Nf1l21; NF-I/A; Nfia; NFI-A; NFI-L; Nuclear factor 1 A-type; nuclear factor 1/A; nuclear factor I A; nuclear factor I/A; TGGCA-binding protein; Unknown (protein for MGC:128467) | |
| Full length human recombinant protein of human CHIA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction