missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NFIA Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596238
This item is not returnable.
View return policy
Description
Immunogen sequence: AYFVHAADSS QSESPSQPSD ADIKDQPENG HLGFQDSFVT SGVFSVTELV RVSQTPIAAG TGPNFSLSDL ESSSYYSMSP GAMRRSLPST SSTSSTKRLK SVEDEMDSPG EEPFYTGQGR SPGSGSQSSG WHEVEPGMPS PTTLKKSEKS G; Positive Samples: Mouse brain; Cellular Location: Nucleus.
NF-1, also designated CTF, consists of a family of CCAAT box binding proteins that stimulate DNA replication and activate transcription. Analysis of human NF-1 messenger RNA has revealed two forms of the NF-1 protein arising from an alternate splicing of a single NF-1 gene. NF-1 binds its consensus DNA element as a homodimer via an amino-terminal DNA binding domain, and activates transcription through a putatively novel, proline-rich, carboxy terminal transactivation domain. The NF-1 protein has been shown to recognize and bind the adenovirus type 2 promoter and activate transcription of herpes simplex virus thymidine kinase genes. The NF-1 consensus element has been found in the upstream promoter region of myriad eukaryotic genes, including that of Ha-Ras, alpha-globin, HSP 70, GRP 78, Histone H1, myelin basic protein and in the Xenopus laevis vitellogenin gene promoter.
Specifications
| NFIA | |
| Polyclonal | |
| Unconjugated | |
| NFIA | |
| 1110047K16Rik; 9430022M17Rik; CCAAT-box-binding transcription factor; CTF; KIAA1439; NF1A; NF1-A; Nf1l21; NF-I/A; Nfia; NFI-A; NFI-L; Nuclear factor 1 A-type; nuclear factor 1/A; nuclear factor I A; nuclear factor I/A; TGGCA-binding protein; Unknown (protein for MGC:128467) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18027, 25492, 4774 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P09414, Q02780, Q12857 | |
| NFIA | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction