Learn More
NFIB Antibody (S03-0E5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19716-100ul
 View more versions of this product
Product Code. 18657943
  • € 645.00 / 100µL
Description
NFIB Monoclonal antibody specifically detects NFIB in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NFIB
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Rat
Purified
Western Blot
S03-0E5
Unconjugated
CCAAT-box-binding transcription factor, CTF, HMGIC/NFIB, NF-I/B, NFI-B, NFIB2, NFIB3, NFI-RED, nuclear factor 1 B-type, Nuclear factor 1/B, nuclear factor I/BNF1-B, TGGCA-binding protein
A synthetic peptide of human NFIB (Uniprot # O00712)
100 μg
Cell Cycle and Replication
4781
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.