Learn More
NFIL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11773-1-AP-20UL
 View more versions of this product
Product Code. 16819073
  • € 145.00 / 20µL
Description
MCM2 (Mini chromosome maintece protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. Mincheva et al. (1994) mapped the gene to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).
Specifications
Specifications
NFIL3
Polyclonal
Unconjugated
NFIL3
E4 promoter binding protein 4, E4BP4, IL3BP1, NF IL3A, NFIL3, NFIL3A
Rabbit
Antigen Affinity Chromatography
RUO
114519, 18030, 4783
-20°C
Liquid
Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O08750, Q16649, Q6IMZ0
Nfil3
NFIL3 Fusion Protein Ag2380
20 μL
Primary
Human, Rat, Mouse