missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ NFkB p105/p50 Transcription Factor Assay Kit (Colorimetric)
Shop All Bio Techne ProductsSpecifications
Specifications
| Product Type | Transcription Factor Assay Kit (Colorimetric) |
| Sample Type | Nuclear Extracts |
| For Use With (Application) | Functional |
| Gene ID (Entrez) | 4790 |
| Gene Symbol | NFKB1 |
| Quantity | 1 Kit |
| Research Discipline | Apoptosis, Cancer, Cell Biology, Diabetes Research, DNA Repair, Neurodegeneration, Neuroscience, Nucleotide Excision Repair, Signal Transduction, Transcription Factors and Regulators |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Synonym | DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?