Learn More
Novus Biologicals™ NFkB p50 (NLS) Inhibitor Peptide Set
Product Code. p-5538750 Shop All Bio Techne Products
Click to view available options
Quantity:
2 mg
5 mg
Unit Size:
2mg
5mg
Product Code. 18705373
Brand: Novus Biologicals™ NBP229323
Applications: Functional (Inhibition)
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Quantity 2 mg