missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFkB1/NFkB p105 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77395H
This item is not returnable.
View return policy
Description
NFkB1/NFkB p105 Polyclonal specifically detects NFkB1/NFkB p105 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| NFkB1/NFkB p105 | |
| Polyclonal | |
| Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| P19838 | |
| NFKB1 | |
| A genomic peptide made to an N-terminal region of the human NFkB p105/p50 protein (within residues 400-590). [Swiss-Prot P19838] | |
| Affinity Purified | |
| RUO | |
| 4790 | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| HRP | |
| PBS with No Preservative | |
| DNA-binding factor KBF1, EBP-1, NF-kappa-B, NF-kB1, NFKB1, NFKB-p105, Nuclear factor NF-kappa-B p105 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1, p105 | |
| Rabbit | |
| 105 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction