missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFKBIE Monoclonal Antibody (OTI1E5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810678S
This item is not returnable.
View return policy
Description
NFKBIE Monoclonal antibody specifically detects NFKBIE in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| NFKBIE | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O54946, O75190, Q6AYU3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10049, 23950, 362293 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1E5 | |
| Unconjugated | |
| DNAJB6 | |
| ikappaBepsilon; I-kappa-B-epsilon; IkB epsilon; IKBE; IkB-E; IkBepsilon; ikB-epsilon; NF-kappa-B inhibitor epsilon; NF-kappa-BIE; NFKB inhibitor epsilon; NFKBIE; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, epsilon; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, epsilon; Slc35b2; solute carrier family 35, member B2 | |
| Full length human recombinant protein of human DNAJB6 produced in HEK293T. | |
| 30 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction