Learn More
NFKBIE Monoclonal Antibody (OTI4D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810661
Encompass_Preferred
 View more versions of this product
Product Code. 17544024
  • € 540.87 / 100µL
Description
Inhibits NF-kappa-B by complexing with and trapping it in the cytoplasm. Inhibits DNA-binding of NF-kappa-B p50-p65 and p50-c-Rel complexes.
Specifications
Specifications
NFKBIE
Monoclonal
Unconjugated
NFKBIE
IKBE
Mouse
Affinity Chromatography
RUO
4794
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI4D1
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O00221
NFKBIE
Full length human recombit protein of human NFKBIE produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a