Learn More
NFKBIE Mouse anti-Human, Clone: OTI5D11, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF810764
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Inhibits NF-kappa-B by complexing with and trapping it in the cytoplasm. Inhibits DNA-binding of NF-kappa-B p50-p65 and p50-c-Rel complexes.Specifications
| NFKBIE | |
| Monoclonal | |
| Unconjugated | |
| NFKBIE | |
| IKBE | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4794 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI5D11 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O00221 | |
| NFKBIE | |
| Full length human recombit protein of human NFKBIE produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.