missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFKBIL1 Polyclonal Antibody

NFKBIL1 Polyclonal Antibody
Brand: Invitrogen PA568974
This item is not returnable.
View return policy
Description
NFKBIL1 Polyclonal Antibody for Western Blot
Specifications
| NFKBIL1 | |
| Polyclonal | |
| Unconjugated | |
| NFKBIL1 | |
| DADB-70P7.2; Def-7; ikappaBL; I-kappa-B-like protein; Ikbl; Inhibitor of kappa B-like protein; LST1; NF-kappa-B inhibitor-like protein 1; NFKB inhibitor like 1; NFKBIL; NFKBIL1; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor like 1; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18038 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O88995 | |
| NFKBIL1 | |
| synthetic peptide directed towards the middle region of human NFKBIL1 (aa 320-369). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction