missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFKBIL2 Antibody (648), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-43746
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
NFKBIL2 Monoclonal specifically detects NFKBIL2 in Human samples. It is validated for Western Blot.
Specifications
| NFKBIL2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with No Preservative | |
| TONSL | |
| Recombinant protein encompassing a sequence within the N-terminus region of human NFKBIL2. The exact sequence is proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 648 | |
| Western Blot 1:500-1:3000 | |
| IkappaBR, I-kappa-B-related protein, IKBRFLJ40087, Inhibitor of kappa B-related protein, NF-kappa-B inhibitor-like protein 2, NFKBIL2tonsoku-like protein, Nuclear factor of kappa light polypeptide gene enhancer in B-cellsinhibitor-like 2ikappaBR, tonsoku-like, DNA repair protein | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 4796 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction