missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NFRKB Recombinant Rabbit Monoclonal Antibody (4N8H3)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555502
This item is not returnable.
View return policy
Description
Immunogen Sequence: LGINEISSSF FSLLLEILLL ESQASLPMLE ERVLDWQSSP ASSLNSWFSA APNWAELVLP ALQYLAGESR AVPSSFSPFV EFKEKTQQWK LLGQSQDNEK ELAALFQLWL ETKDQAFCKQ ENEDSS.
NFRKB (nuclear factor related to kappa-B-binding protein) was first identified as a factor that could bind a variant NF-kappa B site in the interleukin-2 receptor alpha (IL-2R alpha) gene. NFRKB appears to be a transcription factor central to immune cell development. Evidence of this comes from the identification of NFRKB as an amplified gene in several cases of acute myeloid leukemia. Additionally, in another clinical case, a partial deletion of the chromosomal region that includes NFRKB is shown to be associated with combined cellular immunodeficiency.
Specifications
| NFRKB | |
| Recombinant Monoclonal | |
| 1.68 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q6P4R8 | |
| NFRKB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 370-495 of human NFRKB. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 4N8H3 | |
| Unconjugated | |
| NFRKB | |
| A530090G11Rik; DNA-binding protein R kappa B; DNA-binding protein R kappa-B; INO80 complex subunit G; INO80G; NFRKB; nuclear factor related to kappa B binding protein; nuclear factor related to kappaB binding protein; nuclear factor related to kappa-B-binding protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4798 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction