missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFS1 Monoclonal Antibody (OTI5D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805820
This item is not returnable.
View return policy
Description
Iron-sulfur clusters are required for the function of many cellular enzymes. The protein encoded by this gene supplies inorganic sulfur to these clusters by removing the sulfur from cysteine, creating alanine in the process. This gene uses alternate in-frame translation initiation sites to generate mitochondrial forms and cytoplasmic/nuclear forms. Selection of the alternative initiation sites is determined by the cytosolic pH. The encoded protein belongs to the class-V family of pyridoxal phosphate-dependent aminotransferases. Two transcript variants encoding two different isoforms have been found for this gene.Specifications
| NFS1 | |
| Monoclonal | |
| Unconjugated | |
| NFS1 | |
| HUSSY-08, IscS, NIFS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9054 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5D1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y697 | |
| NFS1 | |
| Human recombit protein fragment corresponding to amino acids 1-299 of human NFS1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction